Plugs for test tubes
16 mm, rosso
Codice catalogo 212-0520
Nome del fornitore: BOETTGER PAUL
Vai a
Product Details & Documents
Polyethylene (PE). Ribbed plugs with two sealing ribs, very tight sealing.
- Not autoclavable
- Easy handling with a high, ribbed grip
- Easy to open by tilting 'one-hand operation'
Specifiche
- Colore:rosso
- Seq:08
- Taglia:16 mm