Glass - trace elements, NIST standards
NIST-614 glass - trace elements, 1
Catalog # PROONIST-614
Jump to:
Discontinued
Product Details & Documents
This standard reference material is intended to facilitate the development of chemical methods of analysis for trace elements.
Zertifizierung: Traceable to NIST and SRM.
Specifications
Specifications
- Conf:1
- Descrizione:NIST-614 glass - trace elements