Orders LinkContactUsLinkComponent
Glass - trace elements, NIST standards
Glass - trace elements, NIST standards
NIST-612 glass - trace elements, 1
Codice catalogo PROONIST-612
Glass - trace elements, NIST standards
Codice catalogo PROONIST-612
Nome del fornitore:  
Numero CAS:  
Glass - trace elements, NIST standards
NIST-612 glass - trace elements, 1
Numero CAS:  
Discontinued
Product Details & Documents
This standard reference material is intended to facilitate the development of chemical methods of analysis for trace elements.
Certifications: Traceable to NIST and SRM.
Specifiche
Specifiche
  • Conf:
    1
  • Descrizione:
    NIST-612 glass - trace elements