Reserpine 98%
Bottiglia di vetro, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:50-55-5
- Formula:C₃₃H₄₀N₂O₉
- MDL Number:MFCD00005091
- Molecular Weight:608.69 g/mol
- Melting Pt:265 °C (dec.)
- Storage Temperature:Ambiente
- ADR:6.1,III
Specifications
Specification Test Results
- Appearance:White to yellow-brown Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (60°C, 3 hrs)
- Sulfated ash:≤0.1 %
- HPLC:≥97.5 % (on dried basis)
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:1 g



