RapID™ System
RapID™ Ss/U panel, 20
Codice catalogo OXOIR8311004
Nome del fornitore: THERMO FISHER DIAGNOSTICS
RapID™ System
Codice catalogo OXOIR8311004
Nome del fornitore: THERMO FISHER DIAGNOSTICS
Numero CAS:
Vai a
Product Details & Documents
RapID™ Systems are a complete line of cost-effective manual identification panels for a broad spectrum of microorganisms. RapID™ panels offer easy, one-step, simultaneous inoculation of dehydrated reactants in tray wells to identify anaerobes, coryneform Gram-positive bacilli, enterics, nonfermenters, streptococci, and yeast all within 4 hours. Systems are ideal for manual testing or as confirmation for automated test systems.
- Rapid, accurate identification of over 400 medically important organisms
- Simple, one-step inoculation
- Four-hour aerobic incubation
- Easy-to-read colour reactions
Documents
Certificates
Cerca per numero di lottoSpecifiche
Specifiche
- Conf:20
- Descrizione:RapID™ Ss/U panel