RapID™ System
RapID™ CB Plus panel, 20
Codice catalogo OXOIR8311008
Nome del fornitore: THERMO FISHER DIAGNOSTICS
RapID™ System
Codice catalogo OXOIR8311008
Nome del fornitore: THERMO FISHER DIAGNOSTICS
Numero CAS:
Vai a
Product Details & Documents
RapID™ Systems are a complete line of cost-effective manual identification panels for a broad spectrum of microorganisms. RapID™ panels offer easy, one-step, simultaneous inoculation of dehydrated reactants in tray wells to identify anaerobes, coryneform Gram-positive bacilli, enterics, nonfermenters, streptococci, and yeast all within 4 hours. Systems are ideal for manual testing or as confirmation for automated test systems.
- Rapid, accurate identification of over 400 medically important organisms
- Simple, one-step inoculation
- Four-hour aerobic incubation
- Easy-to-read colour reactions
Documents
Certificates
Cerca per numero di lottoSpecifiche
Specifiche
- Conf:20
- Descrizione:RapID™ CB Plus panel
Specifiche
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.