Orders LinkContactUsLinkComponent
Protein reducing agents | Avantor
Protein reducing agents
Protein-S-S-Reductant™, 200 Tests
Codice catalogo 786-25PR
Nome del fornitore:   G-Biosciences
Protein reducing agents
Codice catalogo 786-25PR
Nome del fornitore:  G-Biosciences
Numero CAS:  
Protein reducing agents
Protein reducing agents
Protein-S-S-Reductant™, 200 Tests
Numero CAS:  
Price
SFr. 183.00
Log in per disponibilità
1 * 200 Tests
Product Details & Documents
Protein-S-S-Reductant™ uses TCEP (Tris [2-carboxyethyl] phosphine), a popular alternative to β-mercaptoethanol and DTT (dithiothreitol). TCEP improves stability, increases effectiveness, and reduces proteins over a wider range of pH, including lower acidic pHs. Protein-S-S-Reductant™ completely reduces stable disulfide bonds in less than 5 minutes at room temperature and is compatible with the protein alkylation reactions. This ready to use solution is at a neutral pH and stabilised for long-term storage. Simply supplement Protein-S-S-Reductant™ in place of DTT or β-mercaptoethanol and boil the sample.
OneQuant™ DTT are single aliquots of DTT that eliminate the need for weighing; preventing loss of reagent and saving time. Add 90 μl water to a single tube to generate a 0,5 M DTT solution. Supplied with 40 individual tubes.
Specifiche
Specifiche
  • Conf:
    200 Tests
  • Descrizione:
    Protein-S-S-Reductant™