Orders LinkContactUsLinkComponent
Normal cell line blots, human
Normal cell line blots, human
Normal muscle cell line blot, human, 1
Codice catalogo TB53
Nome del fornitore:   G-Biosciences
Normal cell line blots, human
Codice catalogo TB53
Nome del fornitore:  G-Biosciences
Numero CAS:  
Normal cell line blots, human
Normal cell line blots, human
Normal muscle cell line blot, human, 1
Numero CAS:  
Price
1 170.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Ready to screen, normal tissue human cell line blots from epithelial, endothelial, skin, and muscle tissue.
These premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4 to 20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using western transfer buffer. These blots are ready to be blocked and probed with antibodies of choice.
Specifiche
Specifiche
  • Conf:
    1
  • Descrizione:
    Normal muscle cell line blot, human