Blocking solution, BLOK™ Casein
BLOK™ Casein in PBS, 1% solution, 2
Codice catalogo 786-194
Nome del fornitore: G-Biosciences
Blocking solution, BLOK™ Casein
Codice catalogo 786-194
Nome del fornitore: G-Biosciences
Numero CAS:
Vai a
Product Details & Documents
BLOK™ Casein is a purified protein, used for blocking free binding sites in Western blot, ELISA plate well, immunohistochemistry and other immunochemical applications.
- Ready to use
- Available in the choices of TBS or PBS buffers
Documents
Specifiche
Specifiche
- Conf:2
- Descrizione:BLOK™ Casein in PBS, 1% solution