Orders LinkContactUsLinkComponent
Proteomic protein controls | Avantor
Proteomic protein controls
Protein proteomic control set, 4×2 mg, 1 SET
Codice catalogo PSC-05
Nome del fornitore:   G-Biosciences
Proteomic protein controls
Codice catalogo PSC-05
Nome del fornitore:  G-Biosciences
Numero CAS:  
Proteomic protein controls
Protein proteomic control set, 4×2 mg, 1 SET
Numero CAS:  
Price
SFr. 397.00
Log in per disponibilità
1 * 1 SET
Product Details & Documents
Protein preparations specifically prepared for standardising electrophoresis methods and protocols. Substantially free from non-protein agents such as nucleic acids, detergents, salts, lipids and other common laboratory agents, the protein preparation has low conductivity (<50 μs). Supplied as dry protein pellets, simply re-hydrate and use. Proteomic protein sample controls are available for animal cells, E.coli, yeast, plant or as a control set that contains one 2 mg vial of each.
Specifiche
Specifiche
  • Conf:
    1 SET
  • Descrizione:
    Protein proteomic control set, 4×2 mg
Specifiche
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..