Orders LinkContactUsLinkComponent
RNaseOUT™ RNAse removal solution
RNaseOUT™ RNAse removal solution
RNaseOut™, refill, 1 L
Codice catalogo 786-71
Nome del fornitore:   G-Biosciences
RNaseOUT™ RNAse removal solution
Codice catalogo 786-71
Nome del fornitore:  G-Biosciences
Numero CAS:  
RNaseOUT™ RNAse removal solution
RNaseOUT™ RNAse removal solution
RNaseOut™, refill, 1 L
Numero CAS:  
Price
116.00 SFr.
Log in per disponibilità
1 * 1 L
Product Details & Documents
RNaseOUT™ is uniquely formulated to destroy and remove RNases on contact.
  • Simply spray and rinse
  • RNaseOUT™ is non-toxic and residue free
  • RNaseOUT™ allows common equipment to be safely and confidently used for all RNA work
Specifiche
Specifiche
  • Conf:
    1 L
  • Descrizione:
    RNaseOut™, refill