Staining racks
Rack, for 24 slides, 435×175 mm
N° de catalogue DELTS-002
Fournisseur: DELTALAB
Aller à
Product Details & Documents
Stainless steel staining racks for microscope slides with and without dividers.
- Good chemical resistance
- Good temperature resistance
Documents
Spécifications
- Descrizione:Rack, for 24 slides, 435×175 mm
- Seq:01