Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Bottiglioni

Bottiglioni

5 L, UN approval: UN 3H1/Y/1.9, 150×195×250 mm, GL 51
Codice catalogo KART1781
Nome del fornitore:   KARTELL
Bottiglioni
Codice catalogo KART1781
Nome del fornitore:  KARTELL
Bottiglioni
Bottiglioni

5 L, UN approval: UN 3H1/Y/1.9, 150×195×250 mm, GL 51

Price
SFr. 587.00
Log in per disponibilità
1 * 40 ST
Product Details & Documents

HDPE, translucent with black screw cap.

  • With comfortable handle and secure seal cap following DIN recommendations
  • Conform to UN regulation on transportation of dangerous liquids
  • Suitable for foodstuff

Ideal for storing and transporting liquids.

Specifiche
  • Filettatura:
    GL 51
  • Capacità:
    5 L
  • Descrizione:
    UN approval: UN 3H1/Y/1.9
  • L×P×A:
    150×195×250 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.