Distilling heads
19/26 NS, 24/29 NS
Codice catalogo WITG1.010.004
Nome del fornitore: witeg Labortechnik
Vai a
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With one NS socket and two NS cones, descending angle: 75°.
Specifiche
- Giunto conico per condensatore:19/26 NS
- Giunto conico per matraccio:24/29 NS