Reversed phase bulk packing material
PRP-h5 bulk resin, 12 - 20 µm, 300 Å, 1 g
Codice catalogo 554-1404
Nome del fornitore: HAMILTON BONADUZ
Reversed phase bulk packing material
Codice catalogo 554-1404
Nome del fornitore: HAMILTON BONADUZ
Numero CAS:
Vai a
Product Details & Documents
Polymer bulk packing material for reversed phase applications.
Chromatographic capacity and efficiency tests are conducted on both bulk and packed column resins to ensure product integrity.
Documents
Specifiche
Specifiche
- Dimensioni particelle:12 - 20 µm
- Conf:1 g
- Dimensioni pori:300 Å
- Descrizione:PRP-h5 bulk resin
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



