Accessori
Tappi Cryobank/Bank-It™, rosso
Katalognr. 479-1075
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Ensure contamination-free cap replacement with Thermo Scientific™ Nunc™ Cryobank and Bank-it™ Caps, which are supplied racked for manual or automated replacement.
- Available in different colours for simplified sample tracking
- Racked in 96 microplate pattern for easy contamination free pick
Technische Daten
- Colore:rosso
- Descrizione:Tappi Cryobank/Bank-It™
- Seq:34
- Cat. No.:479-1075
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:IRECS