Orders LinkContactUsLinkComponent
Staining dishes, WHEATON®
Staining dishes, WHEATON®
Staining racks with handle for 50 slides, - mm
Codice catalogo 631-2507
Nome del fornitore:   DWK Life Sciences
Staining dishes, WHEATON®
Codice catalogo 631-2507
Nome del fornitore:  DWK Life Sciences
Staining dishes, WHEATON®
Staining dishes, WHEATON®
Staining racks with handle for 50 slides, - mm
Price
SFr. 363.00
Log in per disponibilità
1 * 3 ST
Product Details & Documents
Soda-lime glass and stainless steel.
  • Handle permanently attached
  • Hinged closure
  • Easy insertion and removal of slides
The slotted rack hold 50 microscope slides 75×25 mm, 75×38 mm or 75×51 mm. Rack is made of non tarnishable, stainless steel that is resistant to staining solutions.
Specifiche
  • Descrizione:
    Staining racks with handle for 50 slides
  • L×P×A:
    - mm
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..