Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Stirrer guides, universal, PTFE

Stirrer guides, universal, PTFE

Stirrer guide, with standard tapper, 24/29 NS, 10 mm, 96 mm
Codice catalogo COWI005.205.7
Nome del fornitore:   COWIE
Stirrer guides, universal, PTFE
Codice catalogo COWI005.205.7
Nome del fornitore:  COWIE
Stirrer guides, universal, PTFE
Stirrer guides, universal, PTFE

Stirrer guide, with standard tapper, 24/29 NS, 10 mm, 96 mm

Price
SFr. 513.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents

Stirrer guide features composite PTFE/PEEK seal and a glass ball-bearing for rigidity and smoothness of operation.

  • Resistant to chemicals
  • Anti-whip and reduced vibration

They have an NPT screw thread fitting for direct assembly into PTFE reactor lids or NPT extension adapters and standard taper cone fittings are for use in standard glass joints.

Specifiche
  • Descrizione:
    Stirrer guide, with standard tapper, 24/29 NS
  • Altezza:
    96 mm
  • Seq:
    13
  • Ø corpo:
    10 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.