Tute, Secutex® pro 5/6
bianco, L
113-0650
This particulate resistant disposable overall made from PP-spun fabric and PE laminate protects against fine dusts and splashes of aggressive, liquid chemicals. The overall comes complete with a protective flap over the zip, elasticated hood, back, sleeves and ankles.
- Generous cut provides excellent freedom of movement, making it extremely comfortable to wear
- Permeable to air and water vapour
- Flexible, lint-free, abrasion resistant and anti-static material
Colour: White.
Type 5/6, EN 1149, Cat. III/CE 01
: CE 0624, Codice III, Tipo 5 + 6.
- Taglia dell'abbigliamento:L
- Colore:bianco
- Seq:02
- Cat. No.:113-0650
- Antistatico:+
- Applicazioni per abiti:Chemical protection|Laboratory|Industrial