Sonde a alta precisione
PT100 blunt, glass coated probe, TPX 330, 0...400 °C, 8×190 mm
Catalog # 620-1405
Supplier: EBRO
Jump to:
Product Details & Documents
Specifications
- Range di misura:0...400 °C
- Descrizione:PT100 blunt, glass coated probe
- Seq:12
- Tipo:TPX 330
- Ø×L:8×190 mm