Orders LinkContactUsLinkComponent
Sonde a alta precisione
PT100 blunt, glass coated probe, TPX 330, 0...400 °C, 8×190 mm
Catalog # 620-1405
Supplier:   EBRO
Sonde a alta precisione
Catalog # 620-1405
Supplier:  EBRO
Sonde a alta precisione
Sonde a alta precisione
PT100 blunt, glass coated probe, TPX 330, 0...400 °C, 8×190 mm
Price
SFr. 552.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Range di misura:
    0...400 °C
  • Descrizione:
    PT100 blunt, glass coated probe
  • Seq:
    12
  • Tipo:
    TPX 330
  • Ø×L:
    8×190 mm