Volumetric flasks, class A, white markings
5 ml, 10/19, 0,02 ± ml
Codice catalogo 612-2903
Nome del fornitore: SciLabware
Vai a
Product Details & Documents
PYREX®, borosilicate glass, amber, with PE stopper.
- Calibrated to the tolerances required by the United States Pharmacopeia (USP), which are tighter than those of ISO/DIN standards
- Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
- White enamel inscriptions, ideal for use with light sensitive solutions
Certifications: BS EN ISO 1042, DIN 12664
Specifiche
- Cono (NS):10/19
- Capacità:5 ml
- Tolleranza:0,02 ± ml