Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
34×80 mm
Codice catalogo 537-0007
Nome del fornitore:   PRAT DUMAS
Extraction thimbles
Codice catalogo 537-0007
Nome del fornitore:  PRAT DUMAS
Extraction thimbles
Extraction thimbles
34×80 mm
Price
130.00 SFr.
Log in per disponibilità
1 * 25 ST
Product Details & Documents
Pure cotton cellulose
  • Neutral and free of fats and grease
  • Moulded in one piece
  • Very resistant to solvents
Applications include extraction of sludges, asphalt, foodstuffs and in the analysis and quality control of powders and dusts.
Specifiche
  • Ø interno×A:
    34×80 mm