Extraction thimbles
80×200 mm
Codice catalogo 537-0014
Nome del fornitore: PRAT DUMAS
Vai a
Product Details & Documents
Pure cotton cellulose
- Neutral and free of fats and grease
- Moulded in one piece
- Very resistant to solvents
Applications include extraction of sludges, asphalt, foodstuffs and in the analysis and quality control of powders and dusts.
Specifiche
- Ø interno×A:80×200 mm