Sample spoons, mini, SP Bel-Art
0,10 ml, PS
Codice catalogo BELAF367210010
Nome del fornitore: Bel-Art Products, a Part of SP
Sample spoons, mini, SP Bel-Art
Codice catalogo BELAF367210010
Nome del fornitore: Bel-Art Products, a Part of SP
Vai a
Product Details & Documents
PS, white.
- Disposable, made from white high-impact PS
- Ideal for collecting specific small volumes of powders and other dry materials
- 0,10 / 0,50 / 1,0 ml models have a rounded bowl for easier emptying of the spoon and a thinner neck that minimises extra material collection
Documents
Certificates
Cerca per numero di lottoSpecifiche
- Capacità:0,10 ml
- Materiale:PS
- Seq:02