Drying chambers
Heated chamber with ¹/₂" thick, clear acrylic cover, 229×267 mm, 115 V, for Freezone® 4.5A, 4.5P, 6, 12 and 18
Codice catalogo LBCP7521000
Nome del fornitore: LABCONCO
Vai a
Product Details & Documents
Specifiche
- Descrizione:Heated chamber with ¹/₂" thick, clear acrylic cover, 229×267 mm, 115 V, for Freezone® 4.5A, 4.5P, 6, 12 and 18
- Seq:04