Orders LinkContactUsLinkComponent
Wound closure system, Autoclip®
Wound closure system, Autoclip®
Autoclip applicator, 9 mm
Codice catalogo HARV34-0554
Nome del fornitore:   HARVARD APPARATUS
Wound closure system, Autoclip®
Codice catalogo HARV34-0554
Nome del fornitore:  HARVARD APPARATUS
Wound closure system, Autoclip®
Autoclip applicator, 9 mm
Price
SFr. 2’650.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
This Autoclip® wound closure system provides a rapid, easy to use method to securely close wounds. When the wound is healed, simply remove the clip using the handy removal forcep.
  • Provides maximum holding force with minimal trauma
Automatic, spring-fed applicators permit rapid, unassisted application.
Informazioni per l'ordine: Starter set includes 100 wound clips, applicator, and removal forcep.
Specifiche
  • Descrizione:
    Autoclip applicator
  • Seq:
    01
  • Taglia:
    9 mm