Wound closure system, Autoclip®
Autoclip applicator, 9 mm
Codice catalogo HARV34-0554
Nome del fornitore: HARVARD APPARATUS
Vai a
Product Details & Documents
This Autoclip® wound closure system provides a rapid, easy to use method to securely close wounds. When the wound is healed, simply remove the clip using the handy removal forcep.
- Provides maximum holding force with minimal trauma
Automatic, spring-fed applicators permit rapid, unassisted application.
Informazioni per l'ordine: Starter set includes 100 wound clips, applicator, and removal forcep.
Specifiche
- Descrizione:Autoclip applicator
- Seq:01
- Taglia:9 mm