Instrument dishes
1000 ml, 160×290×35 mm
Catalog # BURK4230-2100
Supplier: BURKLE
Jump to:
Product Details & Documents
Trays for various uses. In laboratories, doctors’ surgeries, hospitals, pharmacies, the foodstuffs industry - wherever there is a need for clean storage of instruments, equipment, goods and precision parts.
- Melamine, white
- Tray without lid
Lid available as accessory.
Specifications
- Capacità:1000 ml
- L×P×A:160×290×35 mm