RapID™ System
RapID™ One panel, 20
Nome del fornitore: THERMO FISHER DIAGNOSTICS
RapID™ System
Codice catalogo OXOIR8311006
Nome del fornitore: THERMO FISHER DIAGNOSTICS
Numero CAS:
Vai a
Product Details & Documents
RapID™ Systems are a complete line of cost-effective manual identification panels for a broad spectrum of microorganisms. RapID™ panels offer easy, one-step, simultaneous inoculation of dehydrated reactants in tray wells to identify anaerobes, coryneform Gram-positive bacilli, enterics, nonfermenters, streptococci, and yeast all within 4 hours. Systems are ideal for manual testing or as confirmation for automated test systems.
- Rapid, accurate identification of over 400 medically important organisms
- Simple, one-step inoculation
- Four-hour aerobic incubation
- Easy-to-read colour reactions
Certificates
Cerca per numero di lottoSpecifiche
Specifiche
- Conf:20
- Descrizione:RapID™ One panel
Specifiche
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..