Microarray slides, HTA™
3-D amino microarray slides, 1
Catalog # 731-3021
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
The HTA™ 1 can be used for the processing of one sample. The HTA™ 12 are partitioned into 12 flat compartments, each with a printable surface of 6×6 mm and a low well rim of 0,5 mm. Twelve samples can be processed simultaneously.
- Multifunctional 3-D amino, 3-D aldehyde or 3-D epoxy surface functionality
- Low well rim of only 0,5 mm, low auto-fluorescence
- Plastic plates for high-throughput microarrays
Combine the advantages of the universal microplate platform with microarrays for diagnostic applications due to identical well geometry compared with a standard 96-well microplate.
Specifications
- N. di pozzetti:1
- Descrizione:3-D amino microarray slides