Safety funnels
180 mm filling opening, S 56×4
Codice catalogo DUEH5.10.007
Nome del fornitore: DUPERTHAL
Vai a
Product Details & Documents
Stainless steel safety funnels with hinged lid and butress thread S 56×4.
- Overfilling protection
- Lid reduces the escaping vapours
- Flame arrester made from stainless steel (simple to interchange)
Specifiche
- Filettatura:S 56×4
- Descrizione:180 mm filling opening