Watch glasses
Watch glass, glass, with ground edges, 50 mm
Codice catalogo MURRG850/01
Nome del fornitore: MURRAY S
Vai a
Product Details & Documents
Made of glass.
- Good chemical and physical properties
- Use in applications that do not involve high thermal stresses
- Quick and easy to clean
With ground edges for covering beakers.
Specifiche
- Ø:50 mm
- Descrizione:Watch glass, glass, with ground edges
- Seq:01