Dressing jars
200 mm, 200 mm
Catalog # MURRG500/08
Supplier: MURRAY S
Jump to:
Product Details & Documents
Dressing jars, made of moulded glass, round.
- Good chemical resistance
- Lids must be ordered separately
Specifications
- Ø est:200 mm
- Altezza:200 mm
- Seq:07