Dressing jars
150 mm, 200 mm
Codice catalogo MURRG500/06
Nome del fornitore: MURRAY S
Vai a
Product Details & Documents
Dressing jars, made of moulded glass, round.
- Good chemical resistance
- Lids must be ordered separately
Specifiche
- Ø est:150 mm
- Altezza:200 mm
- Seq:06