Ladles
50 ml, 275 mm, 63 mm, Acciaio inossidabile 304
Codice catalogo SAMPA324-50
Nome del fornitore: Sampling Systems
Vai a
Product Details & Documents
These ladles made from hygienic stainless steel are ideal for sampling liquid products. The ladles have a crevice free construction so that they can be easily cleaned between batches.
- Crevice free construction allows easy cleaning between batches
- Satin finish
Specifiche
- Ø:63 mm
- Capacità:50 ml
- Lunghezza:275 mm
- Materiale:Acciaio inossidabile 304
- Seq:01