Soluzioni ISA
Acid buffer for nitrogen oxide ion selective electrode, 475 ml, Ion selective electrode reagents, Bottle, 475 mL
Codice catalogo ORIO956410
Nome del fornitore: Thermo Orion
Vai a
Product Details & Documents
Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.
- Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements
- Ammonia pH-adjusting ISA and low-level ammonia pH-adjusting ISA both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required
- Ammonia pH-adjusting ISA prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results
- Low-level ammonia pH-adjusting ISA is ideal for ammonia samples with a concentration of 10 ppm and below that do not contain metal interferences
Concentration : 10 ppm
For Use With (Equipment) : Ion selective electrode
For Use With (Application) : Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes
Documents
Specifiche
Specifiche
- Tipo di conf.:Bottle
- Conf:475 mL
- Descrizione:Acid buffer for nitrogen oxide ion selective electrode, 475 ml
- Tipo:Ion selective electrode reagents
Specifiche
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



