Filter candles
4, 13×25 mm
Codice catalogo LENZ07206425
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
These cylindrical filter plugs without tube are made of borosilicate glass 3,3.
Documents
BSE TSE Glass Columns
Certificati BSE/TSE
Specifiche
- Ø×A:13×25 mm
- Porosità:4
- Seq:10