Filter candles
3, 9×20 mm
Codice catalogo LENZ07205320
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
These cylindrical filter plugs without tube are made of borosilicate glass 3,3.
Documents
BSE TSE Glass Columns
Certificati BSE/TSE
Specifiche
- Ø×A:9×20 mm
- Porosità:3
- Seq:04