Flange clamps
Sfera e flange piatte KF/PF 15
Codice catalogo 201-0120
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
Plastic, black.
- Easy to assemble and disassemble
- Tough construction, good temperature resistance
- Matching gaskets are available as an option
For fast, secure connection of ball or flat flanges. No gasket.
Specifiche
- Per:Sfera e flange piatte KF/PF 15
- Seq:01