Inserti di estrazione
Extraction insert for solids, for extractor 250 ml capacity (500 ml volume)
Codice catalogo LENZ05431203
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
DURAN®, Borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
Glass insert with bores for solids.
Documents
BSE TSE Glass Columns
Certificati BSE/TSE
Specifiche
- Descrizione:Extraction insert for solids, for extractor 250 ml capacity (500 ml volume)
- Seq:12