Cold traps with/without condensation drain
1000 ml, without condensate drain
Codice catalogo LENZ05355070
Nome del fornitore: Lenz Laborglas
Cold traps with/without condensation drain
Codice catalogo LENZ05355070
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
For direct filling with coolant. Double-walled design ensures low coolant consumption. Includes NS 29/32 standard ground socket for connecting to the apparatus and NS 29/32 standard ground cone for connecting to the vacuum pump.
DURAN®, chiaro.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Specifiche
- Descrizione:1000 ml, without condensate drain
- Seq:03