Orders LinkContactUsLinkComponent
Cold traps with/without condensation drain
Cold traps with/without condensation drain
500 ml, without condensate drain
Codice catalogo LENZ05355058
Nome del fornitore:   Lenz Laborglas
Cold traps with/without condensation drain
Codice catalogo LENZ05355058
Nome del fornitore:  Lenz Laborglas
Cold traps with/without condensation drain
Cold traps with/without condensation drain
500 ml, without condensate drain
Price
878.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
For direct filling with coolant. Double-walled design ensures low coolant consumption. Includes NS 29/32 standard ground socket for connecting to the apparatus and NS 29/32 standard ground cone for connecting to the vacuum pump.
DURAN®, chiaro.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Documents
Specifiche
  • Descrizione:
    500 ml, without condensate drain
  • Seq:
    02