Condensatori, Davies
400 mm, 29/32 NS, 29/32 NS
Codice catalogo LENZ05215414
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
DURAN® borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With double sized coiling area. Water connection with two GL 14 threaded plastic olives.
Documents
BSE TSE Glass Columns
Certificati BSE/TSE
Specifiche
- Dimensione cono:29/32 NS
- Lunghezza mantello:400 mm
- Cono femmina:29/32 NS