Orders LinkContactUsLinkComponent
Distillation columns, Hempel
800 mm, 29/32 NS, 29/32 NS
  LENZ05117804
Distillation columns, Hempel
  LENZ05117804
Distillation columns, Hempel
Distillation columns, Hempel
800 mm, 29/32 NS, 29/32 NS
SFr. 238.00
1 * 1 ST
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours, with cooling coil and cooling mantle. Standard ground cone and jacket. Water supplied through plastic connectors with 8 mm external Ø and GL threaded joint.
DIN 12593
 : DIN 12593
  • Dimensione cono:
    29/32 NS
  • Cono femmina:
    29/32 NS
  • Lunghezza utile:
    800 mm