Distillation columns, Hempel
500 mm, 29/32 NS, 29/32 NS
LENZ05117504
DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours, with cooling coil and cooling mantle. Standard ground cone and jacket. Water supplied through plastic connectors with 8 mm external Ø and GL threaded joint.
DIN 12593
: DIN 12593
- Dimensione cono:29/32 NS
- Cono femmina:29/32 NS
- Lunghezza utile:500 mm