Pressure bulbs
Mono-ball with two valves, for vacuum, rubber, Ø 46 mm, rosso
Catalog # KART3911
Supplier: KARTELL
Jump to:
Product Details & Documents
Made of rubber, red.
Mono-balls with valve are suitable for pellet burette charging or for vapourisers.
Double balls with net are suitable for automatic burettes and flash chromatography columns.
Tube length: 33,5 cm
Specifications
- Colore:rosso
- Descrizione:Mono-ball with two valves, for vacuum, rubber, Ø 46 mm
- Seq:02