Nitrogen receivers (schlenk Tubes)
250 ml, 14/23 NS, - NS
Codice catalogo LENZ03370249
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
DURAN®.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With 14/23 standard ground sockets/cones and standard ground side stopcock with tube connector.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifiche
- Dimensione cono:14/23 NS
- Cono femmina:- NS
- Capacità:250 ml