Sleeve samplers, U-D Master
Camera di campionamento (tipo 2), 1,3 – 2,5 ml
Codice catalogo SAMP1500A-04
Nome del fornitore: Sampling Systems
Vai a
Product Details & Documents
The U-D sleeve sampler was developed for accurate unit dose samples (small volume).
- Quick and easy accurate unit dose sampling
- Different volumes can be taken using the same sampler
- Simply change the tip for a different volume
Designed for use with free flowing powders and granules.
Specifiche
- Volume:1,3 – 2,5 ml
- Descrizione:Camera di campionamento (tipo 2)
- Seq:05