Orders LinkContactUsLinkComponent
Sleeve samplers, U-D Master
Sleeve samplers, U-D Master
Manico con barra a T, 600 mm
Codice catalogo SAMP1500A-01
Nome del fornitore:   Sampling Systems
Sleeve samplers, U-D Master
Codice catalogo SAMP1500A-01
Nome del fornitore:  Sampling Systems
Sleeve samplers, U-D Master
Sleeve samplers, U-D Master
Manico con barra a T, 600 mm
Price
251.00 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
The U-D sleeve sampler was developed for accurate unit dose samples (small volume).
  • Quick and easy accurate unit dose sampling
  • Different volumes can be taken using the same sampler
  • Simply change the tip for a different volume
Designed for use with free flowing powders and granules.
Specifiche
  • Descrizione:
    Manico con barra a T
  • Lunghezza:
    600 mm
  • Seq:
    02