Orders LinkContactUsLinkComponent
Stoppers for conical ground joints
Stoppers for conical ground joints
19/26, Clear glass
Codice catalogo 217-9098
Nome del fornitore:   Lenz Laborglas
Stoppers for conical ground joints
Codice catalogo 217-9098
Nome del fornitore:  Lenz Laborglas
Stoppers for conical ground joints
Stoppers for conical ground joints
19/26, Clear glass
Price
SFr. 211.00
Log in per disponibilità
1 * 10 ST
Product Details & Documents
DURAN®, borosilicate glass stopper, clear, solid or semi hollow.
  • Excellent chemical resistance
  • High thermal resistance and minimal heat expansion results in high resistance to temperature changes
  • Octagonal top allows easy release and removal of the stopper
Certifications: DIN 12252
Documents
Specifiche
  • Cono (NS):
    19/26
  • Tipo:
    Clear glass