Stoppers for conical ground joints
19/26, Clear glass
Codice catalogo 217-9098
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
DURAN®, borosilicate glass stopper, clear, solid or semi hollow.
- Excellent chemical resistance
- High thermal resistance and minimal heat expansion results in high resistance to temperature changes
- Octagonal top allows easy release and removal of the stopper
Certifications: DIN 12252
Documents
BSE TSE Glass Columns
Certificati BSE/TSE
Specifiche
- Cono (NS):19/26
- Tipo:Clear glass