Single channel pipettes, variable volume, Pluripet
Product Details & Documents
These variable pipettes are equipped with an accurate and precise pipetting mechanism. They cover the range from 0,2 to 10000 µl.
- Special handle ensures a good grip and prevents hand fatigue
- Volume can be selected by turning the emptying button or the cylinder
- Height adjustable tip ejector houses a large variety of tips and can be disassembled to facilitate cleaning operations
- Colour-coded buttons for easy identification
- Robust PVDF body ensures high mechanical and chemical resistance
- Completely autoclavable at +121 °C for 20 minutes
The PL2 and PL10 models are ideal for molecular biology techniques. The PL20, PL100, PL200 and PL1000 models are useful for dispensing small volumes of aqueous fluids of moderate density and viscosity. The PL5000 and PL10000 macropipettes are ideal for dispensing large volumes of rather thick and viscous aqueous fluids. They are equipped with a special filter that protects the internal piston by increasing its lifetime and preventing contaminations.
Specifiche
- Volume:0,5 - 10 µl
- Descrizione:Pluripet PL 10
- Accuratezza:±4,0; ±1,0; ±0,8
- Per puntali:10 µl